Assay Details
Gene Symbol: CHCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046159.2 | NP_001039624.1 | 1 - 2 | 194 | 74 | |
GenBank mRNA | BC112827.1 | - | 1 - 2 | 195 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112827.1, NM_001046159.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
