Assay Details
Gene Symbol: HBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046196.2 | NP_001039661.2 | 7 - 8 | 978 | 86 | |
GenBank mRNA | BC105542.1 | - | 7 - 8 | 967 | 86 | |
BT030675.1 | - | 6 - 7 | 943 | 86 | ||
DV934925.1 | - | 2 - 3 | 185 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
