Assay Details
Gene Symbol: GOLT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046215.2 | NP_001039680.1 | 2 - 3 | 228 | 127 | |
GenBank mRNA | BC110261.1 | - | 2 - 3 | 229 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
