Assay Details
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046230.1 | NP_001039695.1 | 1 - 2 | 86 | 89 | |
GenBank mRNA | BC110249.1 | - | 1 - 2 | 86 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
