Assay Details
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046230.1 | NP_001039695.1 | 2 - 3 | 137 | 88 | |
GenBank mRNA | BC110249.1 | - | 2 - 3 | 137 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
