Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046238.3 | NP_001039703.2 | 21 - 22 | 2939 | 144 | |
GenBank mRNA | BC151637.1 | - | 21 - 22 | 2940 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
