Assay Details
Gene Symbol: LITAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046252.2 | NP_001039717.1 | 1 - 2 | 60 | 84 | |
GenBank mRNA | BC112870.1 | - | 1 - 2 | 62 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112870.1, NM_001046252.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
