Assay Details
Gene Symbol: PSMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046365.2 | NP_001039830.1 | 6 - 7 | 722 | 87 | |
GenBank mRNA | BC111179.1 | - | 6 - 7 | 723 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
