Assay Details
Gene Symbol: DUSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046452.2 | NP_001039917.1 | 3 - 4 | 975 | 72 | |
GenBank mRNA | BC105384.1 | - | 3 - 4 | 976 | 72 | |
DQ473440.1 | - | 3 - 4 | 450 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
