Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046499.2 | NP_001039964.1 | 10 - 11 | 1267 | 112 | |
GenBank mRNA | BC112704.1 | - | 10 - 11 | 1270 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
