Assay Details
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046543.2 | NP_001040008.1 | 1 - 2 | 122 | 66 | |
GenBank mRNA | BC112607.1 | - | 1 - 2 | 123 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112607.1, NM_001046543.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
