Assay Details
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046543.2 | NP_001040008.1 | 3 - 4 | 281 | 93 | |
GenBank mRNA | BC112607.1 | - | 3 - 4 | 282 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
