Assay Details
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046568.1 | NP_001040033.1 | 2 - 3 | 267 | 151 | |
GenBank mRNA | BC111674.1 | - | 2 - 3 | 267 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
