Assay Details
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046594.1 | NP_001040059.1 | 1 - 2 | 173 | 153 | |
GenBank mRNA | BC112771.1 | - | 1 - 2 | 173 | 153 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112771.1, NM_001046594.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
