Assay Details
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046594.1 | NP_001040059.1 | 4 - 5 | 502 | 155 | |
GenBank mRNA | BC112771.1 | - | 4 - 5 | 502 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
