Assay Details
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075166.1 | NP_001068634.1 | 4 - 5 | 721 | 86 | |
GenBank mRNA | BC109588.1 | - | 4 - 5 | 721 | 86 | |
EU005652.1 | - | 4 - 5 | 744 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
