Assay Details
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075797.3 | NP_001069265.2 | 2 - 3 | 203 | 86 | |
GenBank mRNA | BC109850.1 | - | 2 - 3 | 202 | 86 | |
EV618408.1 | - | 2 - 3 | 204 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109850.1, NM_001075797.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
