Assay Details
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075866.2 | NP_001069334.1 | 1 - 2 | 52 | 82 | |
GenBank mRNA | BC112612.1 | - | 1 - 2 | 53 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112612.1, NM_001075866.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
