Assay Details
Gene Symbol: DNAJC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075866.2 | NP_001069334.1 | 3 - 4 | 321 | 79 | |
GenBank mRNA | BC112612.1 | - | 3 - 4 | 322 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112612.1, NM_001075866.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
