Assay Details
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076285.2 | NP_001069753.2 | 4 - 5 | 494 | 100 | |
NM_001113257.2 | NP_001106728.1 | 2 - 3 | 239 | 100 | ||
GenBank mRNA | BC109793.1 | - | 4 - 5 | 456 | 100 | |
EE334754.1 | - | 2 - 3 | 239 | 100 | ||
EV609191.1 | - | 4 - 5 | 497 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
