Assay Details
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076301.2 | NP_001069769.1 | 3 - 4 | 339 | 99 | |
GenBank mRNA | BC109863.1 | - | 3 - 4 | 340 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
