Assay Details
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076379.2 | NP_001069847.1 | 1 - 2 | 139 | 96 | |
GenBank mRNA | BC110187.1 | - | 1 - 2 | 140 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110187.1, NM_001076379.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
