Assay Details
Gene Symbol: DNAJC5G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076429.2 | NP_001069897.1 | 1 - 2 | 209 | 109 | |
GenBank mRNA | BC111333.1 | - | 1 - 2 | 210 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC111333.1, NM_001076429.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
