Assay Details
Gene Symbol: DNAJC5G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076429.2 | NP_001069897.1 | 5 - 6 | 724 | 141 | |
GenBank mRNA | BC111333.1 | - | 5 - 6 | 725 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
