Assay Details
Gene Symbol: FAM228A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077061.2 | NP_001070529.1 | 1 - 2 | 358 | 131 | |
GenBank mRNA | BC109980.1 | - | 1 - 2 | 360 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
