Assay Details
Gene Symbol: FAM228A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077061.2 | NP_001070529.1 | 3 - 4 | 614 | 150 | |
GenBank mRNA | BC109980.1 | - | 3 - 4 | 616 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
