Assay Details
Gene Symbol: COG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077067.2 | NP_001070535.1 | 3 - 4 | 1458 | 134 | |
GenBank mRNA | BC110177.1 | - | 3 - 4 | 1459 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
