Assay Details
Gene Symbol: MMGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079630.1 | NP_001073098.1 | 4 - 5 | 639 | 98 | |
GenBank mRNA | BC109649.1 | - | 4 - 5 | 639 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
