Assay Details
Gene Symbol: VTI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079647.2 | NP_001073115.1 | 5 - 6 | 745 | 86 | |
GenBank mRNA | BC112511.1 | - | 5 - 6 | 746 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
