Assay Details
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079783.1 | NP_001073251.1 | 4 - 5 | 464 | 143 | |
GenBank mRNA | BC111218.1 | - | 4 - 5 | 464 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
