Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079798.1 | NP_001073266.1 | 3 - 4 | 608 | 118 | |
GenBank mRNA | BC109692.1 | - | 3 - 4 | 608 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
