Assay Details
Gene Symbol: ANAPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080357.2 | NP_001073826.1 | 2 - 3 | 584 | 96 | |
GenBank mRNA | BC110224.1 | - | 2 - 3 | 585 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
