Assay Details
Gene Symbol: GNA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174322.4 | NP_776747.1 | 5 - 6 | 979 | 88 | |
GenBank mRNA | BC111118.1 | - | 5 - 6 | 980 | 88 | |
D90336.1 | - | 5 - 6 | 974 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
