Assay Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174788.4 | NP_777213.1 | 77 | 103 | ||
GenBank mRNA | BC110143.1 | - | 78 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110143.1, NM_174788.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
