Assay Details
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044721.1 | NP_001038186.1 | 10 - 11 | 1343 | 84 | |
GenBank mRNA | BC114138.1 | - | 10 - 11 | 1343 | 84 | |
DQ403140.1 | - | 4 - 5 | 563 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
