Assay Details
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044721.1 | NP_001038186.1 | 6 - 7 | 698 | 66 | |
GenBank mRNA | AF461103.1 | - | 3 - 4 | 374 | 66 | |
BC114138.1 | - | 6 - 7 | 698 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
