Assay Details
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045868.1 | NP_001039333.1 | 2 - 3 | 449 | 64 | |
GenBank mRNA | AJ420928.1 | - | 2 - 3 | 400 | 64 | |
BC105484.1 | - | 2 - 3 | 449 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
