Assay Details
Gene Symbol: CMPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046044.1 | NP_001039509.1 | 1 - 2 | 189 | 97 | |
GenBank mRNA | BC112478.1 | - | 1 - 2 | 189 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
