Assay Details
Gene Symbol: HARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046066.2 | NP_001039531.1 | 2 - 3 | 205 | 89 | |
GenBank mRNA | BC112731.1 | - | 2 - 3 | 206 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
