Assay Details
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046068.2 | NP_001039533.1 | 3 - 4 | 365 | 157 | |
GenBank mRNA | BC105368.1 | - | 3 - 4 | 366 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
