Assay Details
Gene Symbol: NDUFAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046105.2 | NP_001039570.1 | 1 - 2 | 102 | 97 | |
GenBank mRNA | BC105350.1 | - | 1 - 2 | 103 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
