Assay Details
Gene Symbol: EIF2S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046117.2 | NP_001039582.1 | 6 - 7 | 650 | 76 | |
GenBank mRNA | BC112875.1 | - | 6 - 7 | 651 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
