Assay Details
Gene Symbol: RHOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046138.2 | NP_001039603.1 | 3 - 4 | 329 | 107 | |
GenBank mRNA | BC114859.1 | - | 3 - 4 | 330 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
