Assay Details
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046151.2 | NP_001039616.1 | 2 - 3 | 178 | 73 | |
GenBank mRNA | BC112509.1 | - | 2 - 3 | 183 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
