Assay Details
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046151.2 | NP_001039616.1 | 4 - 5 | 452 | 127 | |
GenBank mRNA | BC112509.1 | - | 4 - 5 | 457 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
