Assay Details
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046152.2 | NP_001039617.1 | 6 - 7 | 701 | 78 | |
GenBank mRNA | BC113231.1 | - | 6 - 7 | 597 | 78 | |
EH153594.1 | - | 6 - 7 | 701 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
