Assay Details
Gene Symbol: SPON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046157.1 | NP_001039622.1 | 1 - 2 | 172 | 139 | |
GenBank mRNA | BC113291.1 | - | 1 - 2 | 172 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC113291.1, NM_001046157.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
