Assay Details
Gene Symbol: SPON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046157.1 | NP_001039622.1 | 3 - 4 | 620 | 139 | |
GenBank mRNA | BC113291.1 | - | 3 - 4 | 620 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
