Assay Details
Gene Symbol: VPS37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046161.2 | NP_001039626.1 | 10 - 11 | 1386 | 129 | |
GenBank mRNA | BC114056.1 | - | 10 - 11 | 1387 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
