Assay Details
Gene Symbol: VPS37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046161.2 | NP_001039626.1 | 11 - 12 | 1461 | 73 | |
GenBank mRNA | BC114056.1 | - | 11 - 12 | 1462 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
